Learn More
Kingfisher Biotech Inc Equine IL-15 ELISA
SDP
Supplier:  Kingfisher Biotech Inc DIY0703E004
Encompass_Preferred
The Equine IL-15 ELISA contains capture antibody standard and detection antibody for development of a Equine IL-15 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers concentrations incubation times incubation temperatures and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include IL15 IL-15 interleukin 15. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5190