Learn More
Kingfisher Biotech Inc Equine IL-17A ELISA
SDP
Supplier:  Kingfisher Biotech Inc DIY0704E003
Encompass_Preferred
The Equine IL-17A ELISA contains capture antibody standard and detection antibody for development of a Equine IL-17A ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers concentrations incubation times incubation temperatures and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include IL17A CTLA8 IL-17 IL-17A IL17 CTLA-8 interleukin 17A. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5191