Learn More
Raybiotech Inc Equine IL-1 alpha (IL-1 F1) ELISA Kit
Supplier:  Raybiotech Inc ELEIL1A1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.82 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Equine IL-1 alpha (IL-1 F1)
Catalog No. 50-194-8905