Learn More
Raybiotech Inc Equine IL-1 Ra (IL-1 F3) ELISA Kit
Supplier:  Raybiotech Inc ELEIL1RA1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 10 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The sandwich ELISA antibody pair detects Equine IL-1RA.
Catalog No. 50-194-8906