Learn More
Kingfisher Biotech Inc Equine IL-5 Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0104E100
Encompass
The Equine IL-5 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Equine IL-5 applications are for cell culture, ELISA standard, and Western Blot Control. The Equine IL-5 yeast-derived recombinant protein can be purchased in multiple sizes. Equine IL-5 Specifications: (Molecular Weight: 12.9 kDa) (Amino Acid Sequence: AVESPMNRLVAETLTLLSTHRTLLIGDGNLMIPTPEHKNHQLCIEEVFQGIDTLKNQTVQGDAVAKLFQNLSLIKGYIDLQKKKCGGERWRVKQFLDYLQEFLGVINTEWTIEG) (Gene ID: 100034199). For research use only. Made in the USA
Catalog No. 50-241-3787