Learn More
Kingfisher Biotech Inc Equine IL-8 (CXCL8) Polyclonal Antibody (100 ug)
SDP
Supplier:  Kingfisher Biotech Inc PB0433E100
Encompass
The Equine IL-8 (CXCL8) polyclonal antibody is unlabeled and has been qualified for use in ELISA and Western blot applications. It is the capture antibody in the Kingfisher Biotech Equine IL-8 ELISA. The Equine IL-8 polyclonal antibody was produced in goats and is antigen-affinity purified. The reactivity by species is: (Bovine IL-8 - None) (Canine IL-8 - None) (Dolphin IL-8 - None) (Equine IL-8 - Strong) (Feline IL-8 - Weak) (Guinea Pig IL-8 - None) (Human IL-8 - None). For research use only. Made in the USA
Catalog No. 50-233-2235