Learn More
Med Vet International Equine Incisor Extraction Kit, J1614, Each
Catalog No. 5000584418
Encompass_Preferred
Especially designed to aid in the extraction of equine incisors. Six different size heads with a curved concave head and sharp pointed edge. Packed in an attractive zipper case. • 3 Instruments: Wider heads and heavier black teflon handle. 20cm (8") L • 3 Instruments: Finer heads and lighter stainless steel shaft. 22cm (9") L • Weight: 2.3 lbs.
Catalog No. 50-005-84418 Supplier Med Vet International Supplier No. JOR1614
Especially designed to aid in the extraction of equine incisors. Six different size heads with a curved concave head and sharp pointed edge. Packed in an attractive zipper case. • 3 Instruments: Wider heads and heavier black teflon handle. 20cm (8") L • 3 Instruments: Finer heads and lighter stainless steel shaft. 22cm (9") L • Weight: 2.3 lbs.