Learn More
Raybiotech Inc Equine MCP-1/CCL2 ELISA Kit
Supplier:  Raybiotech Inc ELEMCP11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 8.2 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Equine MCP-1 (CCL2)
Catalog No. 50-194-8909