Learn More
Raybiotech Inc Equine MMP-9 ELISA Kit
Supplier:  Raybiotech Inc ELEMMP91
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 49 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Equine MMP-9
Catalog No. 50-194-8910