Learn More
Kingfisher Biotech Inc Equine IL-17A ELISpot - Up to two (2) ELISpot assays
SDP
Catalog No. 502891826
Encompass_Preferred
Equine IL-17A ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-289-1826 Supplier Kingfisher Biotech Inc Supplier No. SPOT2437E004
Equine IL-17A ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA