Learn More
Med Vet International Equine Silicone Endotracheal Tube 16mm, J0151XA, Each
Catalog No. 5000583621
Encompass_Preferred
Universal fitting allows connection to most large animal anesthesia machines and ventilators.
Catalog No. 50-005-83621 Supplier Med Vet International Supplier No. JOR151XA
Universal fitting allows connection to most large animal anesthesia machines and ventilators.