Learn More
Med Vet International Equine Silicone Tracheostomy Tube, 25mm, J0692D
Catalog No. 5000582056
Encompass_Preferred
Silicone softens at body temperature for patient comfort. Balloon inflates via one-way valve. The universal attachment fits large animal anesthesia machine and ventilator hoses. Has a wide outer flexible plate for secure attachment (i.e., umbilical tape)
Catalog No. 50-005-82056 Supplier Med Vet International Supplier No. JOR692D
Silicone softens at body temperature for patient comfort. Balloon inflates via one-way valve. The universal attachment fits large animal anesthesia machine and ventilator hoses. Has a wide outer flexible plate for secure attachment (i.e., umbilical tape)