Learn More
Kingfisher Biotech Inc Equine TNF alpha Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0137E005
Encompass
The Equine TNF alpha yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Equine TNF alpha applications are for cell culture, ELISA standard, and Western Blot Control. The Equine TNF alpha yeast-derived recombinant protein can be purchased in multiple sizes. Equine TNF alpha Specifications: (Molecular Weight: 17.1 kDa) (Amino Acid Sequence: LRSSSRTPSD KPVAHVVANP QAEGQLQWLS GRANALLANG VKLTDNQLVV PLDGLYLIYS QVLFKGQGCP STHVLLTHTI SRLAVSYPSK VNLLSAIKSP CHTESPEQAE AKPWYEPIYL GGVFQLEKGD QLSAEINQPN YLDFAESGQV YLGIIAL (157)) (Gene ID: 100033834). For research use only. Made in the USA
Catalog No. 50-241-3678