Learn More
Med Vet International Equine Uterine Lavage Administration Kit, Short 6" Inflow W/ Female Spin Luer Lock Adapter, J1052A
Catalog No. 5000583462
Encompass_Preferred
These large bore tubing kits are designed to connect large Foley catheters for volume infusion and collection. The "Y" junction allows for fluid infusion and outflow for drainage or collection. Sterile.Short 6" inflow with female spin luer lock adapters. This allows hookup to syringe or another administration set. 30" outflow tube. Distal end has a barbed fitting for connection to Foley flushing catheter (JOR282).
Catalog No. 50-005-83462 Supplier Med Vet International Supplier No. JOR1052A
These large bore tubing kits are designed to connect large Foley catheters for volume infusion and collection. The "Y" junction allows for fluid infusion and outflow for drainage or collection. Sterile.Short 6" inflow with female spin luer lock adapters. This allows hookup to syringe or another administration set. 30" outflow tube. Distal end has a barbed fitting for connection to Foley flushing catheter (JOR282).