Learn More
Med Vet International Equine Uterine Lavage Administration Kit Single Line w/ 60" Large Bore Tubing, J1052B
Catalog No. 5000583592
Encompass_Preferred
These large bore tubing kits are designed to connect large Foley catheters for volume infusion and collection. The "Y" junction allows for fluid infusion and outflow for drainage or collection. Sterile.Single line with 60" large bore tubing with proximal spike and distal barbed fitting for attachment to Foley or large bore pipette (JOR286).
Catalog No. 50-005-83592 Supplier Med Vet International Supplier No. JOR1052B
These large bore tubing kits are designed to connect large Foley catheters for volume infusion and collection. The "Y" junction allows for fluid infusion and outflow for drainage or collection. Sterile.Single line with 60" large bore tubing with proximal spike and distal barbed fitting for attachment to Foley or large bore pipette (JOR286).