Learn More
Med Vet International Equine Uterine Lavage "Y" Catheter, 135cm, J0282YL
Catalog No. 5000582087
Encompass_Preferred
"Y" connector, with 12" arms, gives in flow and out flow. Two shut-off clamps. Two different adaptors included.
Catalog No. 50-005-82087 Supplier Med Vet International Supplier No. JOR282YL
"Y" connector, with 12" arms, gives in flow and out flow. Two shut-off clamps. Two different adaptors included.