Learn More
Raybiotech Inc Equine VEGF-A ELISA Kit
Supplier:  Raybiotech Inc ELEVEGFA1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Equine VEGF-A.
Catalog No. 50-194-8914