Learn More
Med Vet International Equine X-ray Cassette Holder, J0296
Catalog No. 5000585167
Encompass_Preferred
A lightweight piece of equipment that allows the cassette to be held at any angle but keeps the person holding it out of the harmful X-ray beam. Will hold both 8" x 10" and 10" x 12" cassettes. 36" shaft. Quick release clamp. Breakdown model for easy storage.
Catalog No. 50-005-85167 Supplier Med Vet International Supplier No. JOR296
A lightweight piece of equipment that allows the cassette to be held at any angle but keeps the person holding it out of the harmful X-ray beam. Will hold both 8" x 10" and 10" x 12" cassettes. 36" shaft. Quick release clamp. Breakdown model for easy storage.