Learn More
Adam Equipment Adam Equipment Equinox Precision Balance EPB 363i 360g capacity x 0.001g readability
Supplier:  Adam Equipment EPB 363I
Encompass_Preferred
Adam Equipment Equinox Precision Balances balances deliver remarkable precision to labs that perform highly detailed testing. The Equinox is ideal for research and quality assurance facilities; science education; precision counting; and production and manufacturing applications needing exceptionally precise results. Twelve precision models offer capacities from 360g to 8200g and readabilities of 1mg and 0.01g. A color touch screen display provides intuitive operation, with icon-driven menus and readily accessible applications and functions. The five-inch screen is generously sized, offering 800 X 480 pixel resolution for excellent visibility. Featuring a high-resolution color touch screen, Equinox precision balances provide intuitive operation and readily accessible functions. Equinox delivers exceptionally precise results for labs, science education and production facilities. AC adapter included
Catalog No. NC2612359