Learn More
MSC EQUIPMENT CLNR/DEGREASR
Catalog No. 502816801
Encompass
This liquid, butyl-free cleaner and degreaser is designed for industrial use, particularly in food and beverage plants. It is a non-acidic, dilutable concentrate, offering an environmentally preferable biodegradable solution for various cleaning needs. It has a low VOC content of 10 g/L.
  • Liquid form
  • Drum container type
  • Container size of 55 gal
  • No scent
  • Not a disinfectant
  • Butyl-free formula
  • Dilutable concentrate solution
  • Ideal for food & beverage plants
  • Non-acidic composition
  • VOC content of 10 g/L
  • Biodegradable for environmental benefits
Catalog No. 50-281-6801 Supplier MSC Supplier No. 33701731
This liquid, butyl-free cleaner and degreaser is designed for industrial use, particularly in food and beverage plants. It is a non-acidic, dilutable concentrate, offering an environmentally preferable biodegradable solution for various cleaning needs. It has a low VOC content of 10 g/L.
  • Liquid form
  • Drum container type
  • Container size of 55 gal
  • No scent
  • Not a disinfectant
  • Butyl-free formula
  • Dilutable concentrate solution
  • Ideal for food & beverage plants
  • Non-acidic composition
  • VOC content of 10 g/L
  • Biodegradable for environmental benefits