Learn More
Electron Microscopy Sciences EMS Equipment Stage 50 mm
SDP
Supplier:  Electron Microscopy Sciences 45001
Encompass_Preferred
These specimen stages are all easy-change, drop-in style (no screws), and are height adjustable. Standard stage with six stub positions for 15 mm or 6.5 mm or 1/8" pin stubs (same as #3330, 6401, 6552). Compatible with all EMS series.
Catalog No. 50-190-2423