Learn More
Electron Microscopy Sciences EMS Equipment Stage 6" Square Wafer
SDP
Supplier:  Electron Microscopy Sciences 450011
Encompass_Preferred
These specimen stages are all easy-change, drop-in style (no screws), and are height adjustable. Stage to accept 6" square wafer or Masks. Compatible with EMS150TT and EMS150RR.
Catalog No. 50-190-2433