Learn More
Electron Microscopy Sciences EMS Equipment Stage 6" Wafer
SDP
Supplier:  Electron Microscopy Sciences 45004
Encompass_Preferred
These specimen stages are all easy-change, drop-in style (no screws), and are height adjustable. Adjusts to accept 4" & 6" wafers comes with 4500-7, a 6" stub holder to accept up to 27 1/8 pin stubs (same as #6547). Compatible with EMS300.
Catalog No. 50-190-2426