Learn More
Electron Microscopy Sciences EMS Equipment Stage Slide
SDP
Supplier:  Electron Microscopy Sciences 450017
Encompass_Preferred
These specimen stages are all easy-change, drop-in style (no screws), and are height adjustable. Microscope slide stage for up to two 75 mm x 25 mm slides or eight stub positions for pin stubs. (same as #3370, 4520, 6403, 6554). Compatible with all EMS series.
Catalog No. 50-190-2439