Learn More
Med Vet International Equivet Magnet Floats, Convex float for occlusive surface
Supplier:  Med Vet International XJOR1048G
Encompass_Preferred
A totally new approach to a float blade attachment to the handle. These floats have a magnetic head that holds securely the solid carbide blade. The blades can be removed with a "T" bar handle punch. The blades are a very sharp solid carbide with rounded corners. Hollow shaft lightens weight. The shaft is flattened at its attachment to the handle. This gives the operator orientation on the blade location. Limited Supply. No backorders. No Returns
Catalog No. 50-005-84099