Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (±)-Equol-d4 | 1216469-13-4 | MFCD34567407 | 99.9% | 246.29 g/mol | C15H10D4O3 | 5 MG
SDP

Catalog No. 5000262803
Encompass_Preferred

(±)-Equol-d4 is a deuterium-labeled analogue of (±)-equol supplied for research use as a stable isotope tracer and internal standard in quantitative and analytical assays. The compound is characterized by high isotopic and chemical purity and is provided in small lab-friendly pack sizes for analytical workflows.

  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • High purity (99.93%) suitable for analytical quantitation.
  • Molecular formula C15H10D4O3; molecular weight 246.29 g/mol.
  • Available in small pack sizes convenient for analytical workflows.
  • Soluble in common organic solvents such as DMSO for sample preparation.
  • Intended for research use only; not for human therapeutic use.

Catalog No. 50-002-62803 Supplier Medchemexpress LLC Supplier No. HY132421S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(±)-Equol-d4 is a deuterium-labeled analogue of (±)-equol supplied for research use as a stable isotope tracer and internal standard in quantitative and analytical assays. The compound is characterized by high isotopic and chemical purity and is provided in small lab-friendly pack sizes for analytical workflows.

  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • High purity (99.93%) suitable for analytical quantitation.
  • Molecular formula C15H10D4O3; molecular weight 246.29 g/mol.
  • Available in small pack sizes convenient for analytical workflows.
  • Soluble in common organic solvents such as DMSO for sample preparation.
  • Intended for research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.