Learn More
Abcam Equol, Metabolite of phytoestrogen daidzein, 5MG
Catalog No. 502707620
Encompass_Preferred
MW 242.27 Da, Purity >98%. Isoflavan metabolite produced by the reduction of daidzein (ab120391) by gut micro-organisms. Shown to have estrogenic activity in human breast cancer cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7620 Supplier Abcam Supplier No. AB1203905MG
MW 242.27 Da, Purity >98%. Isoflavan metabolite produced by the reduction of daidzein (ab120391) by gut micro-organisms. Shown to have estrogenic activity in human breast cancer cells.
The product is subject to the following: Abcam Restricted Use Statement