Learn More
Medchemexpress LLC HY-100852 1mg Medchemexpress, ER-000444793 CAS:792957-74-5 Purity:>98%
SDP
Catalog No. 501931971
Encompass_Preferred
Medchemexpress, HY-100852 1mg ER-000444793 CAS:792957-74-5 ER-000444793 is a potent inhibitor of mitochondrial permeability transition pore ( mPTP ) opening. ER-000444793 inhibits mPTP with an IC 50 of 2.8 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1971 Supplier Medchemexpress LLC Supplier No. HY1008521MG
Medchemexpress, HY-100852 1mg ER-000444793 CAS:792957-74-5 ER-000444793 is a potent inhibitor of mitochondrial permeability transition pore ( mPTP ) opening. ER-000444793 inhibits mPTP with an IC 50 of 2.8 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.