Learn More
Biocare Medical, LLC Estrogen Receptor (ER) [1D5] Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC ACA054C
Encompass_Preferred
The estrogen receptor antibody (ER) is a 66 kDa protein that acts as an estrogen-dependent, nuclear hormone receptor. Studies have shown ER is present in the nuclei of epithelial cells in normal breast and endometrial tissues, as well as a subset of breast carcinomas. The ER protein has six functionally discrete domains; labeled A through F. ER [1D5] reacts with the amino-terminal domain in the A/B region of ER-alpha. This clone has been established to work in formalin-fixed, paraffin-embedded tissues and has been published in numerous breast cancer research studies.
Catalog No. 50-824-58