Learn More
Medchemexpress LLC ER-27319 maleate | 1204480-26-1 | 99.4% | 396.44 | 100 MG
SDP
Catalog No. 5000345205
Encompass_Preferred
ER-27319 maleate is an acridone derivative, a potent and selective SYK inhibitor that inhibits the tyrosine phosphorylation of SYK and its activity. It inhibits the release of antigen-induced allergic mediators from human and rat mast cells with an IC50 of 10 μM, making it suitable for the study of allergic diseases.
  • Inhibits antigen-induced generation of inositol phosphates
  • Inhibits release of arachidonic acid
  • Inhibits secretion of histamine and tumor necrosis factor α
  • Selectively inhibits the tyrosine phosphorylation of SYK
Catalog No. 50-003-45205 Supplier Medchemexpress LLC Supplier No. HY108489100MG
ER-27319 maleate is an acridone derivative, a potent and selective SYK inhibitor that inhibits the tyrosine phosphorylation of SYK and its activity. It inhibits the release of antigen-induced allergic mediators from human and rat mast cells with an IC50 of 10 μM, making it suitable for the study of allergic diseases.
  • Inhibits antigen-induced generation of inositol phosphates
  • Inhibits release of arachidonic acid
  • Inhibits secretion of histamine and tumor necrosis factor α
  • Selectively inhibits the tyrosine phosphorylation of SYK