Learn More
Medchemexpress LLC ER-27319 (maleate) | 1204480-26-1 | 99.4% | 396.44 | 50 MG
SDP
Catalog No. 5000346423
Encompass_Preferred
ER-27319 (maleate), an acridone derivative, is a potent and selective SYK inhibitor. It inhibits the tyrosine phosphorylation of SYK and its activity, and can be used for study in allergic diseases. It also inhibits the release of antigen-induced allergic mediators from human and rat mast cells with an IC50 of 10 μM.
  • Potent and selective SYK inhibitor
  • Inhibits tyrosine phosphorylation of SYK
  • Inhibits release of antigen-induced allergic mediators
  • Suitable for study in allergic diseases
Catalog No. 50-003-46423 Supplier Medchemexpress LLC Supplier No. HY10848950MG
ER-27319 (maleate), an acridone derivative, is a potent and selective SYK inhibitor. It inhibits the tyrosine phosphorylation of SYK and its activity, and can be used for study in allergic diseases. It also inhibits the release of antigen-induced allergic mediators from human and rat mast cells with an IC50 of 10 μM.
  • Potent and selective SYK inhibitor
  • Inhibits tyrosine phosphorylation of SYK
  • Inhibits release of antigen-induced allergic mediators
  • Suitable for study in allergic diseases