Learn More
Medchemexpress LLC ER-27319 maleate | 1204480-26-1 | 99.4% | 396.44 g/mol | C22H24N2O5 | 5 MG
SDP
Catalog No. 5000263787
Encompass_Preferred
ER-27319 maleate is an acridone-derivative small molecule that acts as a potent, selective spleen tyrosine kinase (Syk) inhibitor. It inhibits Syk tyrosine phosphorylation and activity, and suppresses antigen-induced release of allergic mediators from human and rat mast cells (reported IC50 ≈ 10 μM). The compound is supplied as the maleate salt in solid form for research use.
  • Potent, selective Syk inhibitor.
  • Inhibits tyrosine phosphorylation of Syk.
  • Suppresses antigen-induced mediator release from mast cells (IC50 ≈ 10 μM).
  • Provided as the maleate salt in solid form for laboratory use.
  • Characterized molecular properties: C22H24N2O5, MW 396.44 g/mol.
  • Intended for research applications in Syk-mediated signaling and allergy models.
Catalog No. 50-002-63787 Supplier Medchemexpress LLC Supplier No. HY1084895MG
ER-27319 maleate is an acridone-derivative small molecule that acts as a potent, selective spleen tyrosine kinase (Syk) inhibitor. It inhibits Syk tyrosine phosphorylation and activity, and suppresses antigen-induced release of allergic mediators from human and rat mast cells (reported IC50 ≈ 10 μM). The compound is supplied as the maleate salt in solid form for research use.
  • Potent, selective Syk inhibitor.
  • Inhibits tyrosine phosphorylation of Syk.
  • Suppresses antigen-induced mediator release from mast cells (IC50 ≈ 10 μM).
  • Provided as the maleate salt in solid form for laboratory use.
  • Characterized molecular properties: C22H24N2O5, MW 396.44 g/mol.
  • Intended for research applications in Syk-mediated signaling and allergy models.