Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Er-464195-01 | 859218-37-4 | 99.7% | 379.02 g/mol | C23H39ClN2 | 10MG
SDP

Catalog No. 5000236357
Encompass_Preferred

ER-464195-01 is an orally active small-molecule inhibitor that disrupts calreticulin binding to integrin α subunits and is provided as a high-purity solid for preclinical in vitro and in vivo research. The compound is DMSO-soluble and has defined storage recommendations for both powder and solution forms.

  • Inhibits calreticulin-integrin α subunit interaction.
  • High purity: 99.7%.
  • Chemical formula: C23H39ClN2.
  • Molecular weight: 379.02 g/mol.
  • Appearance: white to off-white solid.
  • Solubility: soluble in DMSO (≈5 mg/mL) with ultrasonic warming.
  • Storage: powder at -20°C (long term) or 4°C; solutions at -80°C or -20°C.

Catalog No. 50-002-36357 Supplier Medchemexpress LLC Supplier No. HY12468610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ER-464195-01 is an orally active small-molecule inhibitor that disrupts calreticulin binding to integrin α subunits and is provided as a high-purity solid for preclinical in vitro and in vivo research. The compound is DMSO-soluble and has defined storage recommendations for both powder and solution forms.

  • Inhibits calreticulin-integrin α subunit interaction.
  • High purity: 99.7%.
  • Chemical formula: C23H39ClN2.
  • Molecular weight: 379.02 g/mol.
  • Appearance: white to off-white solid.
  • Solubility: soluble in DMSO (≈5 mg/mL) with ultrasonic warming.
  • Storage: powder at -20°C (long term) or 4°C; solutions at -80°C or -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.