Learn More
Genetex Inc Estrogen Receptor alpha Monoclonal Antibody [MSVA-564R] HistoMAX™ (GTX04474) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04474
Encompass_Preferred
The Estrogen Receptor alpha antibody [MSVA-564R] HistoMAX™ (GTX04474) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human endometrium (proliferation) tissue using GTX04474 Estrogen Receptor alpha antibody [MSVA-564R] HistoMAX. Among normal tissues highest levels of estrogen receptor alpha immunostaining occur in epithelial and stromal cells of the endometrium.
Catalog No. NC3667778