Learn More
Enzo Life Sciences Estrogen receptor alpha monoclonal antibody (33) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX803004C050
Encompass_Preferred
Clone: 33. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 247-261 (E247VGMMKGGIRKDRRG261) of the DNA binding domain of ERα (estrogen receptor α). This sequence is completely conserved in many species including mouse, rat, pig, bovine, sheep, chicken and frog. Species reactivity: Human, Mouse, Rat, Monkey, Bovine. Specificity: Recognizes both the steroid occupied and unoccupied forms of ERα. Applications: Flow Cytometry, ICC, IF, IHC (FS), IHC (PS), IP, WB. Formulation: Liquid. In PBS, containing 0.05% sodium azide, 1mg/mL BSA and 30% glycerol.
Catalog No. 50-200-9725