Learn More
Enzo Life Sciences Estrogen receptor beta polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ALX210132C100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 512-530 (C512SSTEDSKN KESSQNLQSQ530) of C-terminal rat ERβ (estrogen receptor β). Species reactivity: Human, Mouse, Rat. Applications: ICC, IF, IHC (FS), IHC (PS), WB. Purity: Epitope-affinity purified. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8073