Learn More
Enzo Life Sciences Estrogen receptor beta polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ALX210178C050
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 467-485 (C467CSTEDSKNKEGSQNLQSQ485) of C-terminal mouse ERβ (estrogen receptor β, isoform β-6). Species reactivity: Human, Rat. Applications: IF, IHC (PS), WB. Purity: Epitope-affinity purified. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-200-8086