Learn More
Enzo Life Sciences Estrogen receptor beta polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ALX210135C050
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 100-115 (A100EPQKSPWCEARSLEH115) of human, mouse and rat ERβ (estrogen receptor β). UniProt ID: Q92731. Species reactivity: Human, Mouse, Rat, Sheep, Monkey. Creossreactivity: Does not cross-react with ERα. Applications: ICC, IHC (FS), IHC (PS), WB Gel Shift. Recommended Dilutions: Gel Shift Assay (1-5µg/ml) Immunocytochemistry (2-4µg/ml) Immunohistochemistry (paraffin sections, 1-2µg/ml) Western Blot (1-2µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Epitope-affinity purified IgG. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8075