Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Estrogen Receptor beta (ER beta) ER Beta Recombinant Protein

Catalog No. A15664
Encompass_Preferred
Change view
Click to view available options
Quantity:
180 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15664 180 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15664 Supplier Invitrogen™ Supplier No. A15664
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. ER beta full-length recombinant protein is: Flexible—use in your in-house assay or as part of our PolarScreen™ ER Beta Competitive Binding Assay for a complete assay solution Ready to use—active, purified, and native (untagged) Convenient—available in bulk sizes ER beta recombinant protein is manufactured under the strictest quality parameters. This product is guaranteed for 6 months after purchase. Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond. Protein Form: Recombinant, Full Length.

Estrogen Receptor beta (ER beta, NR3A2) protein is approximately 55kD, though multiple isoforms ranging from 36-59kD have been described. Human ER beta was initially cloned and characterized from testis. Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Estrogen receptor beta (ER beta) binds estrogens with an affinity similar to that of estrogen receptor alpha. Some ER beta isoforms dominantly inhibit the activity of estrogen receptor alpha in reproductive tissues. ER beta is found widely in many tissues throughout the body and can act as a potent tumor suppressor, playing a crucial role in many cancer types such as prostate cancer, autism spectrum disorder, medullary thyroid carcinoma, De Quervain Disease and risk of cardiovascular disease.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92731
Concentration See Label
For Use With (Application) Kinase Assay
Formulation Proprietary buffer with no preservative
Gene ID (Entrez) 2100
Molecular Weight (g/mol) 53.4 kDa
Name Estrogen Receptor beta (ER beta) ER Beta
Purification Method Purified
Quantity 180 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias ER; ER beta; ER[b]; Erb; Erb2; Erbeta; ER-beta; Esr2; ESRB; ESR-BETA; ESTRB; estrogen receptor 2; estrogen receptor 2 (beta); estrogen receptor 2 (ER beta); estrogen receptor 2 beta; estrogen receptor beta; estrogen receptor beta 1 isoform deletion variant; estrogen receptor beta 4; NR3A2; Nuclear receptor subfamily 3 group A member 2; oestrogen receptor beta; steroid receptor
Common Name Estrogen Receptor beta
Gene Symbol ESR2
Protein Length 477
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag Untagged
Sequence 477
Content And Storage Store at -68 to -85°C.
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.