Learn More
Cell Signaling Technology ER Homeostasis Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. 502725432
Encompass
The ER Homeostasis Antibody Sampler Kit provides an economical solution for detecting proteins involved in endoplasmic reticulum (ER) homeostasis, including those that regulate ER stress and ER-phagy. Each kit contains a carefully selected panel of primary antibodies and a secondary antibody, providing sufficient material to perform two western blot experiments with each primary antibody.
  • Includes antibodies for monitoring key ER stress markers and ER-phagy cargo receptors.
  • Supports analysis of proteins such as BiP, IRE1α, PERK, ATF-4, and XBP-1s.
  • Supplied with an HRP-linked anti-rabbit IgG secondary antibody for detection.
  • Optimized for western blotting applications.
  • Suitable for use in research studying metabolic and neurological disorders, as well as cancer.
Catalog No. 50-272-5432 Supplier Cell Signaling Technology Supplier No. 53898T
The ER Homeostasis Antibody Sampler Kit provides an economical solution for detecting proteins involved in endoplasmic reticulum (ER) homeostasis, including those that regulate ER stress and ER-phagy. Each kit contains a carefully selected panel of primary antibodies and a secondary antibody, providing sufficient material to perform two western blot experiments with each primary antibody.
  • Includes antibodies for monitoring key ER stress markers and ER-phagy cargo receptors.
  • Supports analysis of proteins such as BiP, IRE1α, PERK, ATF-4, and XBP-1s.
  • Supplied with an HRP-linked anti-rabbit IgG secondary antibody for detection.
  • Optimized for western blotting applications.
  • Suitable for use in research studying metabolic and neurological disorders, as well as cancer.