Learn More
Salimetrics LLC Salivary Cortisol Elisa Kit
Supplier:  Salimetrics LLC 130025
Encompass
The Salimetrics Cortisol Enzyme Immunoassay Kit is a competitive immunoassay specifically designed and validated for the quantitative measurement of salivary cortisol. It is not intended for diagnostic use. It is intended only for research use in humans and some animals. Salimetrics has not validated this kit for serum or plasma samples.
Catalog No. NC1833143