Learn More
Enzo Life Sciences ER-ID Red assay kit (GFP-CERTIFIED) (1x1Kit)
SDP
Catalog No. NC0075338
Encompass_Preferred
Endoplasmic  Reticulum staining with minimal toxicity to cells. Applications: Fluorescence microscopy. Contents: ER-ID Red detection reagent: 50μl. Hoechst 33342 nuclear stain: 50μl. 10X assay buffer: 15ml
Catalog No. NC0075338 Supplier Enzo Life Sciences Supplier No. ENZ51026K500
Endoplasmic  Reticulum staining with minimal toxicity to cells. Applications: Fluorescence microscopy. Contents: ER-ID Red detection reagent: 50μl. Hoechst 33342 nuclear stain: 50μl. 10X assay buffer: 15ml