Learn More
LGC GENOMICS LLC (Lucigen) ER2738 Electrocompetent Cells, 12 Rxns (DUOs)Create large phage display libraries with high efficiency cells recommended for peptide display.
Catalog No. NC0292392
Encompass
Perfect for antibody phage display or peptide phage display library creation. ER2738 Electrocompetent Cells are an amber suppressor strain (glnV) prepared as highly efficient (2 10^10 cfu/g) electrocompetent cells for phage display library screening. This strain is recommended for use with New England Biolab's Ph.D.™ Phage Display Kits.
This item is not returnable. View return policy
Catalog No. NC0292392 Supplier LGC GENOMICS LLC Supplier No. 605221
Perfect for antibody phage display or peptide phage display library creation. ER2738 Electrocompetent Cells are an amber suppressor strain (glnV) prepared as highly efficient (2 10^10 cfu/g) electrocompetent cells for phage display library screening. This strain is recommended for use with New England Biolab's Ph.D.™ Phage Display Kits.