Learn More
AMID BIOSCIENCES Amid Biosciences, ER2738 Electrocompetent E. coli Cells, Phage Display, 10 x 0.05 mL
SDP
Catalog No. NC4043476
Encompass_Preferred
ER2738 electrocompetent E. coli cells are supplied in a 10 x 0.05 mL format for phage display library screening and peptide display workflows. The strain is optimized for high efficiency transformation and M13 phage propagation. Applications include phage display, peptide display libraries, M13 phage propagation, high-efficiency library transformation. Technical specifications are included below.- Quantity - 10 x 0.05 mL (20 reactions)- Organism - E. coli- Storage - -80 C- Cell Strain / Host - ER2738- Cell Format - 10 x 0.05 mL electrocompetent cells- Transformation Efficiency - 1.00E+10This product is for research use only.
Catalog No. NC4043476 Supplier AMID BIOSCIENCES Supplier No. EREC20110
ER2738 electrocompetent E. coli cells are supplied in a 10 x 0.05 mL format for phage display library screening and peptide display workflows. The strain is optimized for high efficiency transformation and M13 phage propagation. Applications include phage display, peptide display libraries, M13 phage propagation, high-efficiency library transformation. Technical specifications are included below.- Quantity - 10 x 0.05 mL (20 reactions)- Organism - E. coli- Storage - -80 C- Cell Strain / Host - ER2738- Cell Format - 10 x 0.05 mL electrocompetent cells- Transformation Efficiency - 1.00E+10This product is for research use only.