Learn More
INTACT GENOMICS INC ig ER2738 Phage Display Chemically Competent Cells 24x50μl
Catalog No. 5000082907
Encompass
Intact Genomics (ig) ER2738 Phage Display Chemically Competent Cells are suitable for protein expression and preparation of antibody or peptide phage display libraries. ER2738 cells are also useful for protein expression, M13 phage work, general cloning, and blue/white screening.Product Includes & Storage:ig ER2738 Phage Display Chemically Competent cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4 ºC
Catalog No. 50-000-82907 Supplier INTACT GENOMICS INC Supplier No. 100724
Intact Genomics (ig) ER2738 Phage Display Chemically Competent Cells are suitable for protein expression and preparation of antibody or peptide phage display libraries. ER2738 cells are also useful for protein expression, M13 phage work, general cloning, and blue/white screening.Product Includes & Storage:ig ER2738 Phage Display Chemically Competent cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4 ºC