Learn More
INTACT GENOMICS INC ig ER2738 Phage Display ElectroCompetent Cells 12x100µl
Supplier: INTACT GENOMICS INC 121748
Intact Genomics (ig) ER2738 Phage Display Electrocompetent Cells are suitable for protein expression and preparation of antibody or peptide phage display libraries. ER2738 cells are also useful for protein expression, M13 phage work, general cloning, and blue/ white screening.ig ER2738 Phage Display Electrocompetent Cells have the following features: • >4 x 1010cfu/µg efficiency with electroporation. • Amber suppressor strain (glnV)Product Include and Storage:ig ER2738 Phage Display Electrocompetent Cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4 ºC