Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cambridge Diagnostic Products, Inc.™ Erado-Sol™ Biological and Chemical Stain Remover

Catalog No. 23564105
Change view
Click to view available options
Volume (English):
128 oz. (1 gal.)
16 oz.
2 oz.
32 oz.
8 oz.
Form:
Clear Liquid
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Form
23-564-105 128 oz. (1 gal.) Clear Liquid
23-564-100 - -
23-564-101 2 oz. Clear Liquid
23-564-102 8 oz. Clear Liquid
23-564-103 16 oz. Clear Liquid
23-564-104 32 oz. Clear Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Do not mix with any chemicals other than water. Do not mix with household bleach (Sodium Hypochlorite).
Catalog No. 23-564-105 Supplier Cambridge Diagnostic Products, Inc.™ Supplier No. 307G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Do not mix with any chemicals other than water. Do not mix with household bleach (Sodium Hypochlorite).

Fast and effective biological and chemical stain remover. Erado-Sol Stain Remover helps remove most medical, histological and chemical stains including iodine, Wright Stain, Lugol's Stain and fresh blood stains.

  • Biological and Chemical Stain Remover
  • Fast-Acting
  • Readily Biodegradable Surfactants
  • Neutral pH 7-8
  • No Bleach, Peroxide or Phosphates

Specifications

Product Type Biological and Chemical Stain Remover
Form Clear Liquid
pH 7 to 8
Content And Storage Containers which are open must be carefully resealed and kept upright to prevent leakage
For Use With (Application) Removing stains from hands, glassware, lab equipment, stainless steel, plastic, counters, floors, lab coats, clothing and hospital linens
Volume (English) 128 oz. (1 gal.)
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.