Learn More
Medchemexpress LLC Eragidomide | 1860875-51-9 | 99.9% | 250 MG
SDP
Catalog No. 5000341507
Encompass_Preferred
Eragidomide (CC-90009) is a first-in-class GSPT1-selective cereblon (CRBN) E3 ligase modulator that acts as a molecular glue. It coopts the CRL4CRBN to selectively target GSPT1 for ubiquitination and proteasomal degradation. This product is for research use only and not sold to patients.
  • First-in-class GSPT1-selective cereblon E3 ligase modulator
  • Acts as a molecular glue
  • Coopts the CRL4CRBN to selectively target GSPT1 for ubiquitination and proteasomal degradation
  • For research use only
Catalog No. 50-003-41507 Supplier Medchemexpress LLC Supplier No. 250MGHY130800
Eragidomide (CC-90009) is a first-in-class GSPT1-selective cereblon (CRBN) E3 ligase modulator that acts as a molecular glue. It coopts the CRL4CRBN to selectively target GSPT1 for ubiquitination and proteasomal degradation. This product is for research use only and not sold to patients.
  • First-in-class GSPT1-selective cereblon E3 ligase modulator
  • Acts as a molecular glue
  • Coopts the CRL4CRBN to selectively target GSPT1 for ubiquitination and proteasomal degradation
  • For research use only