Learn More
Aviva Systems Biology Corp ERAL1 Antibody - C-terminal region (ARP40599_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40599_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ERAL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ERAL1. Alternate names: ERA, H-ERA, ERAL1A, HERA-A, HERA-BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57541