Learn More
Aviva Systems Biology Corp ERAS Antibody - middle region (ARP55794_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55794_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ERAS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ERAS. Alternate names: HRAS2, HRASP, MGC126691, MGC126693ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-61616